2,3,5,6-Tetrafluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A102116-100G A102116-25G A102116-5G
77245-166EA 25 USD
77245-166 77245-168 77245-170
2,3,5,6-Tetrafluorobenzyl alcohol 98%
2,3,5,6-Tetrafluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₄F₄O
MDL Number: MFCD00792428
CAS Number: 4084-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR