N,N'-(1,4-Phenylene)diisonicotinamide 97%

Supplier: Ambeed
Warning

A102006-100MG A102006-1G A102006-250MG A102006-5G
77245-158EA 30.9 USD
77245-158 77245-160 77245-162 77245-164
N,N'-(1,4-Phenylene)diisonicotinamide 97%
N,N'-(1,4-Phenylene)diisonicotinamide

Precaución: for Research Use Only

Formula: C₁₈H₁₄N₄O₂
MW: 318.32 g/mol
MDL Number: MFCD00726741
CAS Number: 55119-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR