Monastrol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylic Acid Ethyl Ester, 1,2,3,4-Tetrahydro-4-(3-hydroxyphenyl)-6-methyl-2-thioxo-5-pyrimidinecarboxylic acid, ethyl ester, Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylate, Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, Eg5 Inhibitor I

A101991-10MG A101991-1MG A101991-25MG A101991-50MG A101991-5MG
77257-258EA 19 USD
77257-258 77257-260 77257-262 77257-264 77257-266
Monastrol 98%
Monastrol

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₃S
MW: 292.353 g/mol
MDL Number: MFCD00813077
CAS Number: 329689-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR