CC-401 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(2-(Piperidin-1-yl)ethoxy)phenyl)-5-(1H-1,2,4-triazol-5-yl)-1H-indazole hydrochloride, CC-401 HCl, CC401

A101734-2MG A101734-5MG
77233-088EA 31.93 USD
77233-088 77745-776
CC-401 hydrochloride 98%
CC-401 hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₆O·HCl
MW: 424.9 g/mol
MDL Number: MFCD21364765
CAS Number: 1438391-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR