1,1-Bis((2R,5R)-2,5-di-i-propylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%

Supplier: Ambeed
Warning

A101471-100MG A101471-1G A101471-250MG
77245-130EA 139.05 USD
77245-130 77245-132 77245-134
1,1-Bis((2R,5R)-2,5-di-i-propylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
1,1-Bis((2R,5R)-2,5-di-i-propylphospholano)ferrocene(cyclooctadiene)rhodium(I)

Precaución: for Research Use Only

Formula: C₃₈H₅₀FeP₂Rh
MW: 727.50 g/mol
MDL Number: MFCD15144871
CAS Number: 849773-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR