4-(1H-1,2,4-Triazol-1-yl)aniline 98%

Supplier: Ambeed
Warning

A101440-1G A101440-5G
77245-120EA 15 USD
77245-120 77245-122
4-(1H-1,2,4-Triazol-1-yl)aniline 98%
4-(1H-1,2,4-Triazol-1-yl)aniline

Precaución: for Research Use Only

Formula: C₈H₈N₄
MDL Number: MFCD00171470
CAS Number: 6523-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR