3,4,5-Tribromopyridazine 97%

Supplier: Ambeed
Warning

A101303-100MG A101303-10G A101303-1G A101303-250MG
77245-110EA 5.15 USD
77245-110 77245-112 77245-114 77245-116
3,4,5-Tribromopyridazine 97%
3,4,5-Tribromopyridazine

Precaución: for Research Use Only

Formula: C₄HBr₃N₂
MW: 316.78 g/mol
MDL Number: MFCD00233941
CAS Number: 1822854-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR