8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A101232-100MG A101232-1G A101232-250MG
77257-230EA 40.17 USD
77257-230 77257-232 77257-234
8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%
8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD11847631
CAS Number: 1277178-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR