1-Bromo-4-(bromomethyl)-2-nitrobenzene 95%

Supplier: Ambeed
Danger

A100888-1G A100888-250MG
77282-594EA 20 USD
77282-594 77282-596
1-Bromo-4-(bromomethyl)-2-nitrobenzene 95%
1-Bromo-4-(bromomethyl)-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MDL Number: MFCD08059131
CAS Number: 326595-66-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR