4-(Hexyloxy)-2,3,6-trimethylphenol 98%

Supplier: Ambeed
Warning

Synonyms: HTHQ, HX-1171, 1-O-Hexyl-2,3,5-trimethylhydroquinone

A100799-100MG A100799-10MG A100799-50MG A100799-5MG
77257-216EA 72 USD
77257-216 77257-218 77257-220 77257-222
4-(Hexyloxy)-2,3,6-trimethylphenol 98%
4-(Hexyloxy)-2,3,6-trimethylphenol

Precaución: for Research Use Only

Formula: C₁₅H₂₄O₂
MW: 236.36 g/mol
MDL Number: MFCD00797612
CAS Number: 148081-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR