2-Carboxyethyl acrylate 97% stabilised

Supplier: Ambeed
Danger

Synonyms: 2-Carboxyethylacrylate, 3-Acryloyloxypropionic acid

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A100771-25G A100771-100G
77257-214EA 7 USD
77257-214 77747-753
2-Carboxyethyl acrylate 97% stabilised
2-Carboxyethyl acrylate

Precaución: for Research Use Only

Formula: C₆H₈o₄
MW: 144.126 g/mol
MDL Number: MFCD00040709
CAS Number: 24615-84-7
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR