ZINC00881524 98%

Supplier: Ambeed
Danger

Synonyms: N-(4,5-dihydronaphtho[1,2-d]thiazol-2-yl)-2-(3,4-dimethoxyphenyl)acetamide

A100468-10MG A100468-1MG A100468-50MG A100468-25MG A100468-5MG
77257-202EA 149.35 USD
77257-202 77257-204 77257-206 77296-696 77296-698
ZINC00881524 98%
ZINC00881524

Precaución: for Research Use Only

Formula: C₂₁H₂₀N₂O₃S
MW: 380.46 g/mol
MDL Number: MFCD07821731
CAS Number: 557782-81-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR