3-Bromo-1-indanone 98%

Supplier: Ambeed
Warning

A100288-1G A100288-25G A100288-5G
77270-142EA 32.96 USD
77270-142 77270-144 77270-146
3-Bromo-1-indanone 98%
3-Bromo-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.06 g/mol
MDL Number: MFCD09800595
CAS Number: 40774-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR