(1S,1'S)-1,1'-Bis[bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene 98%
Supplier: Ambeed
|
Warning
|
A100267-100MG
A100267-250MG
77245-074EA
455.99
USD
77245-074
77245-076
77245-078
(1S,1'S)-1,1'-Bis[bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene 98%
(1S,1'S)-1,1'-Bis[bis[3,5-bis(trifluoromethyl)phenyl]phosphino]-2,2'-bis[(S)-(dimethylamino)phenylmethyl]ferrocene
Precaución: for Research Use Only
Formula:
C₆₀H₄₂F₂₄FeN₂P₂ |
MDL Number:
MFCD08561144 CAS Number: 849925-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...