3-(1-(Cyclohexyl(methyl)carbamoyl)-1H-imidazol-4-yl)pyridine 1-oxide 98%

Supplier: Ambeed
Warning

Synonyms: BIA 10-2474

A100075-10MG A100075-1MG A100075-25MG A100075-50MG A100075-5MG
77257-190EA 41.2 USD
77257-190 77257-192 77257-194 77257-196 77257-198
3-(1-(Cyclohexyl(methyl)carbamoyl)-1H-imidazol-4-yl)pyridine 1-oxide 98%
3-(1-(Cyclohexyl(methyl)carbamoyl)-1H-imidazol-4-yl)pyridine 1-oxide

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₄O₂
MW: 300.36 g/mol
MDL Number: MFCD30532661
CAS Number: 1233855-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR