NEBNext® UltraShear™ FFPE DNA Library Preparation Kits
Supplier: New England Biolabs (NEB)
E6655L E6655S
77025-082EA 3270 USD
77025-082 77025-090
NEBNext® UltraShear™ FFPE DNA Library Preparation Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext UltraShear further increases library yields and quality, while improving scalability and ease of use.
  • Includes FFPE DNA fragmentation and repair reagents, plus optimized library prep reagents
  • Increased library yields and improved sequencing metrics
  • Improved scalability and ease of use
  • Greater sensitivity of somatic variant calling Automation-friendly workflow
The NEBNext UltraShear FFPE DNA Library Prep Kit includes NEBNext UltraShear, a new solution designed for enzymatic fragmentation of challenging samples; the NEBNext FFPE DNA Repair v2 Mix, an optimized cocktail of enzymes designed to repair FFPE DNA; library prep reagents featuring a new polymerase master mix, and a protocol optimized for FFPE DNA.