VWR® Max Protection High ESD Cleanroom Coveralls
Supplier: VWR International
76944-256 76944-244 76944-248 76944-258 76944-266 76944-254 76944-280 76944-274 76944-250 76944-270 76944-294 76944-242 76944-262 76944-290 76944-286 76944-272 76944-278 76944-288 76944-276 76944-292 76944-268 76944-260 76944-282 76944-264 76944-246 76944-252 76944-284
76944-256CS 132.91 USD
76944-256 76944-244 76944-248 76944-258 76944-266 76944-254 76944-280 76944-274 76944-250 76944-270 76944-294 76944-242 76944-262 76944-290 76944-286 76944-272 76944-278 76944-288 76944-276 76944-292 76944-268 76944-260 76944-282 76944-264 76944-246 76944-252 76944-284
VWR® Max Protection High ESD Cleanroom Coveralls
Disposable Garments
Cleanroom ESD coveralls are available in a variety of protective formats, including garments with attached hood, non-skid boots, or both.
  • Cleanroom coverall designed to provie high ESD properties
  • Lightweight, comfortable fit
VWR® Max Protection coveralls are made from 63 GSM SF and provide high ESD protection. All coveralls feature a front zipper closure, snap release on the leg, and serged seams. Elastic wrists and ankles are provided on garment without attached hood or boots.
Envase: Coveralls are packaged in a case of 25.