di-Potassium hydrogen phosphate ≥98.0%

Supplier: RPI

P41300-1000.0 P41300-10000.0 P41300-12000.0 P41300-25000.0 P41300-3000.0 P41300-500.0 P41300-5000.0
76347-792EA 101.33 USD
76347-792 76347-794 76347-796 76347-798 76347-800 76347-802 76347-804
di-Potassium hydrogen phosphate ≥98.0%
di-Potassium hydrogen phosphate

  • Soluble in water


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: K₂HPO₄
Melting Pt: 340 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5

Order Now

Specification Test Results

Appearance White Powder
Assay ≥ 98.0 %
Chloride (Cl) ≤ 0.003 %
Heavy Metals ≤ 5 ppm
Insoluble Matter ≤ 0.01 %
Iron ≤ 0.001 %
Loss on Drying ≤ 1.0 %
Nitrogen Compound ≤ 0.001 %
pH (5% Solution) 8.5 - 9.6
Sodium (Na) ≤ 0.05 %
Sulfate ≤ 0.005 %
Aerobic Plate Count ≤ 1000 CFU/g
Total Coliforms Negative
Yeast and Mold Count ≤ 100 CFU/g
BSE/TSE Free BSE/TSE Free


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR