Ethyl [1,4'-bipiperidine]-4-carboxylate dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl1,4’-Bipiperidine-4-CarboxylateHydrochloride

A998306-100MG A998306-250MG
77200-540EA 53.56 USD
77200-540 77200-542
Ethyl [1,4'-bipiperidine]-4-carboxylate dihydrochloride 95%
Ethyl [1,4'-bipiperidine]-4-carboxylate dihydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₆Cl₂N₂O₂
MW: 313.26 g/mol
MDL Number: MFCD26793859
CAS Number: 930604-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR