2-Bromo-3-chloro-1,8-naphthyridine 95%

Supplier: Ambeed
Warning

A996057-100MG A996057-250MG
77172-138EA 307.97 USD
77172-138 77172-140
2-Bromo-3-chloro-1,8-naphthyridine 95%
2-Bromo-3-chloro-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.48 g/mol
MDL Number: MFCD28016819
CAS Number: 52626-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR