Ethylenedioxybis(trimethylsilane) 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane

A994415-10G A994415-1G A994415-25G A994415-5G A994415-100G
77200-530EA 15.45 USD
77200-530 77200-532 77200-534 77200-536 77595-608
Ethylenedioxybis(trimethylsilane) 98%
Ethylenedioxybis(trimethylsilane)

Precaución: for Research Use Only

Formula: C₈H₂₂O₂Si₂
MW: 206.43 g/mol
MDL Number: MFCD00009640
CAS Number: 7381-30-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR