Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A994294-100G
A994294-1G
A994294-25G
A994294-5G
77172-130EA
412
USD
77172-130
77172-132
77172-134
77172-136
Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 97%
Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₂ClNO₃ MW: 217.64 g/mol |
MDL Number:
MFCD00858983 CAS Number: 68697-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...