Calcium bis(trifluoromethylsulfonyl)imide 97%

Supplier: Ambeed
Danger

Synonyms: Calcium(II) Bis(trifluoromethanesulphonyl)imide, Calcium bis(trifluoromethanesulphonimide), Bis(trifluoromethanesulphonyl)imide calcium(II) salt, Calcium(II) triflimide, Calcium triflimide

A990662-100MG A990662-10G A990662-1G A990662-250MG A990662-5G A990662-25G
77172-118EA 7 USD
77172-118 77172-120 77172-122 77172-124 77172-126 77569-720
Calcium bis(trifluoromethylsulfonyl)imide 97%
Calcium bis(trifluoromethylsulfonyl)imide

Precaución: For research use only.

Formula: C₄CaF₁₂N₂O₈S₄
MW: 600.35 g/mol
MDL Number: MFCD14156019
CAS Number: 165324-09-4
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR