1,3-Di(2-pyridyl)-1,3-propanedione 98%

Supplier: Ambeed
Warning

Synonyms: Dipicolinoylmethane, 1,3-Di(pyridin-2-yl)propane-1,3-dione

A989738-1G A989738-250MG A989738-5G A989738-25G
77200-504EA 22 USD
77200-504 77200-506 77200-508 77747-740
1,3-Di(2-pyridyl)-1,3-propanedione 98%
1,3-Di(2-pyridyl)-1,3-propanedione

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O₂
MDL Number: MFCD01321157
CAS Number: 10198-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR