5-tert-Butyl-1,3-benzoxazole-2-thiol 97%

Supplier: Ambeed
Warning

Synonyms: 5-(tert-Butyl)benzo[d]oxazole-2-thiol

A989344-100MG A989344-1G A989344-250MG A989344-5G
77200-496EA 15 USD
77200-496 77200-498 77200-500 77200-502
5-tert-Butyl-1,3-benzoxazole-2-thiol 97%
5-tert-Butyl-1,3-benzoxazole-2-thiol

Precaución: for Research Use Only

Formula: C₁₁H₁₃NOS
MDL Number: MFCD09259264
CAS Number: 53146-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR