5-Fluoro-2-iodoanisole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-1-iodo-2-methoxybenzene

A985528-250MG A985528-25G
77200-490EA 18.54 USD
77200-490 77200-492
5-Fluoro-2-iodoanisole 97%
5-Fluoro-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆FIO
MW: 252.025 g/mol
MDL Number: MFCD18396916
CAS Number: 450-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR