DL-Mevalonolactone 97%

Supplier: Ambeed
Warning

Synonyms: (±)-Mevalonolactone, 4-Hydroxy-4-methyltetrahydro-2H-pyran-2-one, β-Hydroxy-beta-methyl-delta-valerolactone, DL-Mevalonic Acid Lactone, Tetrahydro-4-hydroxy-4-methyl-2-pyrone

A982863-1G A982863-5G A982863-250MG
77172-104EA 56.65 USD
77172-104 77172-106 77684-910
DL-Mevalonolactone 97%
DL-Mevalonolactone

Precaución: For research use only.

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00006648
CAS Number: 674-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR