1,4-Diisopropylbenzene 98%

Supplier: Ambeed
Danger

A982129-25G A982129-5G
77200-480EA 15 USD
77200-480 77200-482
1,4-Diisopropylbenzene 98%
1,4-Diisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₈
MW: 162.27
Boiling Pt: 203
Melting Pt: -40
Density: 0.87707
MDL Number: MFCD00008892
CAS Number: 100-18-5
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR