2-Fluoro-1,3,4,5-tetramethylbenzene 95+%

Supplier: Ambeed
Warning

A980084-1G A980084-5G
77172-096EA 45.32 USD
77172-096 77172-098
2-Fluoro-1,3,4,5-tetramethylbenzene 95+%
2-Fluoro-1,3,4,5-tetramethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃F
MW: 152.21 g/mol
MDL Number: MFCD22683873
CAS Number: 319-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR