tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Aminobenzyl)piperazine, N1-BOC protected, tert-Butyl 4-(4-aminobenzyl)piperazine-1-carboxylate
A976208-10G
A976208-1G
A976208-250MG
A976208-25G
A976208-5G
77172-088EA
254.41
USD
77172-088
77172-090
77172-092
77172-094
77684-907
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate 96%
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate
Precaución: For research use only.
Formula:
C₁₆H₂₅N₃O₂ MW: 291.3886 g/mol |
MDL Number:
MFCD06797795 CAS Number: 304897-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...