1-(2-Chloroethyl)naphthalene 95%

Supplier: Ambeed
Warning

A975946-25G A975946-5G
77200-472EA 520 USD
77200-472 77200-474
1-(2-Chloroethyl)naphthalene 95%
1-(2-Chloroethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁Cl
MW: 190.67 g/mol
MDL Number: MFCD01321149
CAS Number: 41332-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR