6-Bromo-1,4-dimethylquinoxaline-2,3(1H,4H)-dione 97%

Supplier: Ambeed
Warning

A975590-100MG A975590-1G A975590-250MG
77200-466EA 24.72 USD
77200-466 77200-468 77200-470
6-Bromo-1,4-dimethylquinoxaline-2,3(1H,4H)-dione 97%
6-Bromo-1,4-dimethylquinoxaline-2,3(1H,4H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.09 g/mol
MDL Number: MFCD00957063
CAS Number: 1770-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR