3-Cyano-5-(trifluoromethyl)benzoic Acid 95%

Supplier: Ambeed
Warning

A973687-100MG A973687-1G A973687-250MG A973687-5G
77172-072EA 13 USD
77172-072 77172-074 77172-076 77172-078
3-Cyano-5-(trifluoromethyl)benzoic Acid 95%
3-Cyano-5-(trifluoromethyl)benzoic Acid

Precaución: for Research Use Only

Formula: C₉H₄F₃NO₂
MDL Number: MFCD16610738
CAS Number: 942077-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR