(1-Benzyl-1H-benzo[d]imidazol-2-yl)methanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A972633-100MG A972633-1G A972633-250MG
77172-064EA 47.38 USD
77172-064 77172-066 77172-068
(1-Benzyl-1H-benzo[d]imidazol-2-yl)methanamine dihydrochloride 97%
(1-Benzyl-1H-benzo[d]imidazol-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₇Cl₂N₃
MW: 310.22 g/mol
MDL Number: MFCD08752574
CAS Number: 20028-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR