(R)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%

Supplier: Ambeed
Warning

A968560-100MG A968560-1G A968560-250MG
77200-456EA 56 USD
77200-456 77200-458 77200-460
(R)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%
(R)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃₂H₁₉N₂O₈P
MDL Number: MFCD08274951
CAS Number: 695162-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR