Methyl 3-bromo-2-(bromomethyl)benzoate 95%

Supplier: Ambeed
Danger

A967769-100MG A967769-1G A967769-250MG A967769-5G A967769-25G
77172-052EA 28 USD
77172-052 77172-054 77172-056 77172-058 77595-556
Methyl 3-bromo-2-(bromomethyl)benzoate 95%
Methyl 3-bromo-2-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₂
MDL Number: MFCD06804569
CAS Number: 337536-14-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR