2-(Oxan-4-yl)acetaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Tetrahydro-2H-pyran-4-yl)acetaldehyde

A965590-5G A965590-1G A965590-100MG
77200-444EA 323 USD
77200-444 77569-690 77747-723
2-(Oxan-4-yl)acetaldehyde 95%
2-(Oxan-4-yl)acetaldehyde

Precaución: For research use only.

Formula: C₇H₁₂O₂
MW: 128.169 g/mol
MDL Number: MFCD03425265
CAS Number: 65626-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR