1-(4-Iodophenyl)piperidin-2-one 98%
Supplier: Ambeed
|
Warning
|
A963960-1G
A963960-250MG
A963960-25G
A963960-5G
77200-430EA
19.57
USD
77200-430
77200-432
77200-434
77200-436
1-(4-Iodophenyl)piperidin-2-one 98%
1-(4-Iodophenyl)piperidin-2-one
Precaución: for Research Use Only
Formula:
C₁₁H₁₂INO |
MDL Number:
MFCD04037128 CAS Number: 385425-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...