1-Decyl-3-methylimidazolium bromide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Decyl-3-methyl-1H-imidazol-3-ium bromide

A962205-100G A962205-25G A962205-5G
77200-414EA 163 USD
77200-414 77200-416 77200-418
1-Decyl-3-methylimidazolium bromide 98%
1-Decyl-3-methylimidazolium bromide

Precaución: for Research Use Only

Formula: C₁₄H₂₇BrN₂
MW: 303.29
MDL Number: MFCD09038832
CAS Number: 188589-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR