1-Bromo-2-chloro-3-iodobenzene 95%

Supplier: Ambeed
Warning

A961452-100MG A961452-1G A961452-250MG A961452-25G A961452-5G
77172-030EA 10.3 USD
77172-030 77172-032 77172-034 77172-036 77595-552
1-Bromo-2-chloro-3-iodobenzene 95%
1-Bromo-2-chloro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
MW: 317.34 g/mol
MDL Number: MFCD18379733
CAS Number: 57012-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR