1,4-Bis(2,2,2-trifluoroethoxy)benzene 99%

Supplier: Ambeed
Warning

A960779-1G A960779-5G
77200-410EA 25 USD
77200-410 77200-412
1,4-Bis(2,2,2-trifluoroethoxy)benzene 99%
1,4-Bis(2,2,2-trifluoroethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₈F₆O₂
MDL Number: MFCD00042272
CAS Number: 66300-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR