4-(1,1-Difluoroethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A959869-100MG A959869-250MG A959869-1G
77172-024EA 64 USD
77172-024 77172-026 77573-898
4-(1,1-Difluoroethyl)benzoic acid 98%
4-(1,1-Difluoroethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD20643344
CAS Number: 55805-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR