Methyl-2-hydroxynicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxynicotinate, 2-Hydroxynicotinic acid methyl ester

A958894-10G A958894-1G A958894-25G A958894-5G
77172-020EA 76 USD
77172-020 77172-022 77893-756 77893-757
Methyl-2-hydroxynicotinate 95%
Methyl-2-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MDL Number: MFCD08275055
CAS Number: 67383-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR