5,6,7,8-Tetrahydroimidazo[1,2-a]pyrimidine 95%

Supplier: Ambeed
Warning

A958186-250MG A958186-1G A958186-100MG
77172-018EA 44 USD
77172-018 77595-548 77893-755
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrimidine 95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MDL Number: MFCD08273903
CAS Number: 67139-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR