11H-Benzo[b]fluoren-11-one 95%

Supplier: Ambeed
Warning

Synonyms: 11H-Benzo(b)fluoren-11-one

A955226-1G A955226-5G
77200-398EA 46.35 USD
77200-398 77200-400
11H-Benzo[b]fluoren-11-one 95%
11H-Benzo[b]fluoren-11-one

Precaución: for Research Use Only

Formula: C₁₇H₁₀O
MW: 230.27 g/mol
MDL Number: MFCD00215960
CAS Number: 3074-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR