3-Fluoro-N-methylaniline 98%

Supplier: Ambeed
Danger

A953961-1G A953961-25G A953961-5G
77200-392EA 11.33 USD
77200-392 77200-394 77200-396
3-Fluoro-N-methylaniline 98%
3-Fluoro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD02683095
CAS Number: 1978-37-6
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR