2,6-Bis(4,5-dihydrooxazol-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A951283-100MG A951283-10G A951283-1G A951283-250MG A951283-5G
77171-994EA 47 USD
77171-994 77171-996 77171-998 77172-000 77172-002
2,6-Bis(4,5-dihydrooxazol-2-yl)pyridine 97%
2,6-Bis(4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₁N₃O₂
MDL Number: MFCD28145681
CAS Number: 165125-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR