1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

A950630-100MG A950630-1G A950630-250MG
77200-386EA 36.05 USD
77200-386 77200-388 77200-390
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
1-Benzyl-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
MDL Number: MFCD00101801
CAS Number: 4332-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR