1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose

A946407-1G A946407-25G A946407-5G A946407-100G
77200-380EA 16.48 USD
77200-380 77200-382 77200-384 77618-610
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose 98%
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₁₁
MW: 477.42 g/mol
MDL Number: MFCD00080781
CAS Number: 10022-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR