2-[(2-Fluorophenyl)amino]aniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N1-(2-Fluorophenyl)benzene-1,2-diamine, 2-Amino-2'-fluorodiphenylamine
A945002-1G
A945002-25G
A945002-5G
77171-988EA
22.66
USD
77171-988
77171-990
77171-992
2-[(2-Fluorophenyl)amino]aniline 98%
2-[(2-Fluorophenyl)amino]aniline
Precaución: for Research Use Only
Formula:
C₁₂H₁₁FN₂ MW: 202.228 g/mol |
MDL Number:
MFCD10567164 CAS Number: 28898-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...